5-Fluoroindole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Fluoro-1H-indole

31192-0010 31192-0050 31192-0250 31192-2500
TS31192-0010EA 59 USD
TS31192-0010 TS31192-0050 TS31192-0250 TS31192-2500
5-Fluoroindole 98%
5-Fluoroindole
Formula: C₈H₆FN
MW: 135.14 g/mol
Melting Pt: 44…48 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00005671
CAS Number: 399-52-0
EINECS: 206-917-4

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 44 to 48 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR