4-Pentyn-2-ol 97+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Pentyn-4-ol

39267-0010 39267-0050 39267-0250
TS39267-0010EA 27 USD
TS39267-0010 TS39267-0050 TS39267-0250
4-Pentyn-2-ol 97+%
4-Pentyn-2-ol
Formula: C₅H₈O
MW: 84.12 g/mol
Boiling Pt: 126 °C (1013 hPa)
Density: 0.896 g/cm³
Flash Pt: 37 °C
Storage Temperature: Ambient
MDL Number: MFCD00004555
CAS Number: 2117-11-5
UN: 1986
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4360 to 1.4380 (20°C, 589 nm)
GC ≥97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR