4-Chloro-1-butene 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Chlorobut-1-ene

46016-0010 46016-0050
TS46016-0010EA 66 USD
TS46016-0010 TS46016-0050
4-Chloro-1-butene 98%
4-Chloro-1-butene
Formula: C₄H₇Cl
MW: 90.55 g/mol
Density: 0.921 g/cm³
Flash Pt: –8 °C
Storage Temperature: Ambient
MDL Number: MFCD00039386
CAS Number: 927-73-1
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4220 to 1.4240 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR