4-Bromo-2-methoxypyridine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
45000-0010
45000-0050
TS45000-0010EA
96
USD
TS45000-0010
TS45000-0050
4-Bromo-2-methoxypyridine 97%
4-Bromo-2-methoxypyridine
Formula:
C₆H₆BrNO Boiling Pt: 87 °C (15 mmHg) Storage Temperature: Refrigerator |
MDL Number:
MFCD08277268 CAS Number: 100367-39-3 |
Specification Test Results
Appearance | Colorless to yellow Low melting solid or liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5540 to 1.5560 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...