3-Methyl-1-butyne 96%

Supplier: Thermo Scientific Chemicals
Danger

39274-0010 39274-0100
TS39274-0010EA 33 USD
TS39274-0010 TS39274-0100
3-Methyl-1-butyne 96%
3-Methyl-1-butyne
Formula: C₅H₈
MW: 68.12 g/mol
Boiling Pt: 28 °C (1013 hPa)
Melting Pt: –90.1 °C
Density: 0.666 g/cm³
Flash Pt: –30 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00039853
CAS Number: 598-23-2
EINECS: 209-925-6
UN: 3295
ADR: 3,I

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3730 to 1.3750 (20°C, 589 nm)
GC ≥95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR