3-Bromopropionic acid 97%

Supplier: Thermo Scientific Chemicals
Danger

32577-0100 32577-1000 32577-5000
TS32577-0100EA 28 USD
TS32577-0100 TS32577-1000 TS32577-5000
3-Bromopropionic acid 97%
3-Bromopropionic acid
Formula: BrH₂CCH₂COOH
Boiling Pt: 140…142 °C (45 mmHg)
Melting Pt: 57…63 °C
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00002763
CAS Number: 590-92-1
EINECS: 209-694-1
UN: 3261
ADR: 8,II
Merck Index: 1301418

Order Now

Specification Test Results

Appearance White to light yellow to orange Powder or crystals or flakes
Infrared spectrum Conforms
Melting point 57 to 63 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR