2-Bromopyrimidine 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
40323-0010
40323-0050
TS40323-0010EA
29
USD
TS40323-0010
TS40323-0050
2-Bromopyrimidine 97+%
2-Bromopyrimidine
Formula:
C₄H₃BrN₂ Boiling Pt: 62…64 °C (1.5 mmHg) Melting Pt: 55…56 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014601 CAS Number: 4595-60-2 |
Specification Test Results
Appearance | Off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 55 to 59 °C |
HPLC | ≥97.0 % |
Specific optical rotation | -11° to -16° (20 °C, 589 nm) (c=4, MeOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...