1-Iodo-3,3,3-trifluoropropane 98%

Supplier: Thermo Scientific Chemicals
Warning

39774-0010 39774-0050
TS39774-0010EA 21 USD
TS39774-0010 TS39774-0050
1-Iodo-3,3,3-trifluoropropane 98%
1-Iodo-3,3,3-trifluoropropane
Formula: C₃H₄F₃I
Boiling Pt: 89…91 °C (760 mmHg)
Density: 1.911 g/cm³
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00038531
CAS Number: 460-37-7

Order Now

Specification Test Results

Appearance Clear colorless to pink Liquid
Infrared spectrum Conforms
Refractive index 1.4190 to 1.4210 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR