1,13-Tridecanedioic acid

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tridecanedioic acid, Undecane-1,11-dicarboxylic acid, 1,11-Undecanedicarboxylic acid

36944-0050 36944-0250 36944-1000
TS36944-0050EA 61 USD
TS36944-0050 TS36944-0250 TS36944-1000
1,13-Tridecanedioic acid
1,13-Tridecanedioic acid
Formula: C₁₃H₂₄O₄
MW: 244.33 g/mol
Melting Pt: 112…114 °C
Storage Temperature: Ambient
MDL Number: MFCD00002740
CAS Number: 505-52-2

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 112 to 116 °C
GC ≥92 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR