(R)-(+)-3-Butyne-2-ol 98%, 98% ee
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(R)-(+)-But-3-yn-2-ol, (R)-But-3-yn-2-ol, (R)-(+)-3-Hydroxy-1-butyne
36630-0010
36630-0050
TS36630-0010EA
138
USD
TS36630-0010
TS36630-0050
(R)-(+)-3-Butyne-2-ol 98%, 98% ee
(R)-(+)-3-Butyne-2-ol
Formula:
C₄H₆O MW: 70.09 g/mol Boiling Pt: 103 Density: 0.89 g/cm³ Flash Pt: 6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00211237 CAS Number: 42969-65-3 UN: 2929 ADR: 6.1 II |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4235 to 1.4255 (20°C, 589 nm) |
GC | ≥97.5 % |
Enantiomeric excess | ≥97.5 % (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...