5-Iodoindole 95%

Supplier: Thermo Scientific Chemicals
Warning

44159-0050 44159-0250
TS44159-0050EA 100 USD
TS44159-0050 TS44159-0250
5-Iodoindole 95%
5-Iodoindole
Formula: C₈H₆IN
Melting Pt: 95…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00220065
CAS Number: 16066-91-4

Order Now

Specification Test Results

Appearance Off-white to light brown Powder
Infrared spectrum Conforms
Melting point 95 to 102 °C
HPLC ≥94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR