Aluminium acetylacetonate 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tris(2,4-pentanedionato-O,O')aluminium, Acetylacetone Aluminum(III) Salt, Al(acac)3, Aluminium 2,4-pentanedionate, Aluminum(III) Acetylacetonate, Aluminium tris(2,4-pentanedionato-O,O'), Aluminum acetylacetonate, Aluminum 2,4-pentanedionate, Tris(2,4-pentanedionato)aluminum(III), Aluminum triacetylacetone

19697-0050 19697-1000 19697-5000
TS19697-0050EA 19 USD
TS19697-0050 TS19697-1000 TS19697-5000
Aluminium acetylacetonate 97%
Aluminium acetylacetonate
Formula: C₁₅H₂₁AlO₆
MW: 324.31 g/mol
Boiling Pt: 315
Melting Pt: 190…195 °C
Storage Temperature: Ambient
MDL Number: MFCD00000013
CAS Number: 13963-57-0
EINECS: 237-741-6
UN: 3467
ADR: 6.1 II

Order Now

Specification Test Results

Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Melting point 190 to 195 °C
Titration Complexometric ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR