Kojic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Hydroxy-2-hydroxymethyl-4H-pyran-4-one, 5-Hydroxy-2-(hydroxymethyl)-4H-pyran-4-one, 5-Hydroxy-2-hydroxymethyl-4-pyrone, 5-Hydroxy-2-hydroxymethyl-γ-pyrone

20522-0050 20522-0250 20522-1000 20522-5000
TS20522-0050EA 105 USD
TS20522-0050 TS20522-0250 TS20522-1000 TS20522-5000
Kojic acid 99%
Kojic acid
Formula: C₆H₆O₄
MW: 142.11 g/mol
Melting Pt: 151…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00006580
CAS Number: 501-30-4
EINECS: 207-922-4
Merck Index: 1305334

Order Now

Specification Test Results

Appearance White to beige to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 151 to 156 °C
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR