5-Cyano-1-pentyne 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Hexynenitrile

30737-0010 30737-0050
TS30737-0010EA 37 USD
TS30737-0010 TS30737-0050
5-Cyano-1-pentyne 98%
5-Cyano-1-pentyne
Formula: C₆H₇N
MW: 93.13 g/mol
Boiling Pt: 115…117 °C
Density: 0.88 g/cm³
Flash Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD00001978
CAS Number: 14918-21-9
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to amber Liquid
Infrared spectrum Conforms
Refractive index 1.4390 to 1.4410 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR