2,4-Dichloro-5-methylpyrimidine 98%

Supplier: Thermo Scientific Chemicals
Danger

39829-0010 39829-0050
TS39829-0010EA 68 USD
TS39829-0010 TS39829-0050
2,4-Dichloro-5-methylpyrimidine 98%
2,4-Dichloro-5-methylpyrimidine
Formula: C₅H₄Cl₂N₂
MW: 163.01 g/mol
Boiling Pt: 109 °C (11 mmHg)
Melting Pt: 26…28 °C
Density: 1.39 g/cm³
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00023197
CAS Number: 1780-31-0
EINECS: 217-227-8
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance Light yellow Low melting solid
Infrared spectrum Conforms
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR