2,3-Dichloropyrazine 98%

Supplier: Thermo Scientific Chemicals
Warning

37704-0050 37704-0250 37704-1000
TS37704-0050EA 91 USD
TS37704-0050 TS37704-0250 TS37704-1000
2,3-Dichloropyrazine 98%
2,3-Dichloropyrazine
Formula: C₄H₂Cl₂N₂
Boiling Pt: 105…107 °C (50 mmHg)
Melting Pt: 22.5…24.5 °C
Density: 1.431 g/cm³
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD00040964
CAS Number: 4858-85-9
EINECS: 225-460-1

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR