2,3-Dichloroquinoxaline 97%

Supplier: Thermo Scientific Chemicals
Danger

33083-0100 33083-0250
TS33083-0100EA 81 USD
TS33083-0100 TS33083-0250
2,3-Dichloroquinoxaline 97%
2,3-Dichloroquinoxaline
Formula: C₈H₄Cl₂N₂
MW: 199.04 g/mol
Boiling Pt: 301 °C (1013 hPa)
Melting Pt: 150…155 °C
MDL Number: MFCD00006720
CAS Number: 2213-63-0
EINECS: 218-667-3
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Off-white to light brown Needles
Infrared spectrum Conforms
Melting point 150 to 155 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR