3-Chloro-3-methyl-1-butyne 96%

Supplier: Thermo Scientific Chemicals
Danger

37801-0010 37801-0050 37801-0250
TS37801-0010EA 39 USD
TS37801-0010 TS37801-0050 TS37801-0250
3-Chloro-3-methyl-1-butyne 96%
3-Chloro-3-methyl-1-butyne
Formula: C₅H₇Cl
MW: 102.56 g/mol
Flash Pt: 4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00190221
CAS Number: 1111-97-3
EINECS: 214-188-9
UN: 1993
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear colorless to straw yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4170 to 1.4190 (20°C, 589 nm)
GC ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR