1,3-Dibromo-2-propanol 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Glycerol alpha,alpha'-Dibromohydrin, 1,3-Dibromopropan-2-ol, alpha,alpha'-Dibromohydrin

40654-0250 40654-1000
TS40654-0250EA 122 USD
TS40654-0250 TS40654-1000
1,3-Dibromo-2-propanol 95%
1,3-Dibromo-2-propanol
Formula: BrH₂CCH(OH)CH₂Br
MW: 217.89 g/mol
Boiling Pt: 89 °C (1013 hPa)
Density: 2.13 g/cm³
Flash Pt: 46 °C
Storage Temperature: Ambient
MDL Number: MFCD00000216
CAS Number: 96-21-9
EINECS: 202-489-8
UN: 1987
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5500 to 1.5530 (20°C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR