11-Bromoundecanoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 11-Bromohendecanoic acid, 11-Bromoundecylic acid

42316-0010 42316-0250 42316-1000
TS42316-0010EA 695 USD
TS42316-0010 TS42316-0250 TS42316-1000
11-Bromoundecanoic acid 98%
11-Bromoundecanoic acid
Formula: BrH₂C(CH₂)₉COOH
MW: 265.19 g/mol
Boiling Pt: 173…174 °C (2 mmHg)
Melting Pt: 48…51 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00002732
CAS Number: 2834-05-1
EINECS: 220-602-9

Order Now

Specification Test Results

Appearance White to beige Crystalline chunks
Infrared spectrum Conforms
GC ≥97.5 %
Titration Argentometric ≥94 % (ex Cl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR