1-Bromo-2-butyne 98%

Supplier: Thermo Scientific Chemicals
Warning

38702-0010 38702-0050 38702-0250
TS38702-0010EA 41 USD
TS38702-0010 TS38702-0050 TS38702-0250
1-Bromo-2-butyne 98%
1-Bromo-2-butyne
Formula: C₄H₅Br
MW: 132.99 g/mol
Boiling Pt: 40…41 °C (760 mmHg)
Density: 1.473 g/cm³
Flash Pt: 36 °C
Storage Temperature: Ambient
MDL Number: MFCD00190233
CAS Number: 3355-28-0
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear yellow-green Liquid
Infrared spectrum Conforms
Refractive index 1.5070 to 1.5090 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR