2-Adamantanone 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Adamantan-2-one

29250-0050 29250-0250 29250-1000
TS29250-0050EA 50 USD
TS29250-0050 TS29250-0250 TS29250-1000
2-Adamantanone 98%
2-Adamantanone
Formula: C₁₀H₁₄O
MW: 150.22 g/mol
Melting Pt: 282…286 °C
Storage Temperature: Ambient
MDL Number: MFCD00074737
CAS Number: 700-58-3
EINECS: 211-847-2

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR