3-Iodothiophene 97%

Supplier: Thermo Scientific Chemicals
Warning

44338-0010 44338-0050
TS44338-0010EA 79 USD
TS44338-0010 TS44338-0050
3-Iodothiophene 97%
3-Iodothiophene
Formula: C₄H₃IS
Boiling Pt: 195 °C (760 mmHg)
Melting Pt: –13 °C
Density: 2.066 g/cm³
Flash Pt: 78 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00037901
CAS Number: 10486-61-0

Order Now

Specification Test Results

Appearance Colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR