3,4-Dibromothiophene 97%

Supplier: Thermo Scientific Chemicals
Warning

26452-0010 26452-0050 26452-0250
TS26452-0010EA 25 USD
TS26452-0010 TS26452-0050 TS26452-0250
3,4-Dibromothiophene 97%
3,4-Dibromothiophene
Formula: C₄H₂Br₂S
Boiling Pt: 221…222 °C (760 mmHg)
Melting Pt: 4…5 °C
Density: 2.13 g/cm³
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005465
CAS Number: 3141-26-2
EINECS: 221-546-8
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow to light brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.6380 to 1.6420 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR