2-(Bromomethyl)naphthalene 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
16256-0050
16256-0250
16256-1000
TS16256-0010EA
26.78
USD
TS16256-0010
TS16256-0050
TS16256-0250
TS16256-1000
2-(Bromomethyl)naphthalene 96%
2-(Bromomethyl)naphthalene
Formula:
C₁₁H₉Br MW: 221.1 g/mol Boiling Pt: 213 °C (100 mmHg) Melting Pt: 52…57 °C Flash Pt: >110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00004123 CAS Number: 939-26-4 EINECS: 213-359-5 UN: 3261 ADR: 8,II |
Specification Test Results
Appearance | White to light yellow to light beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 52 to 57 °C |
GC | ≥95 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...