2-Bromonaphthalene 99%

Supplier: Thermo Scientific Chemicals
Warning

17971-0050 17971-0250 17971-1000
TS17971-0050EA 45 USD
TS17971-0050 TS17971-0250 TS17971-1000
2-Bromonaphthalene 99%
2-Bromonaphthalene
Formula: C₁₀H₇Br
MW: 207.07 g/mol
Boiling Pt: 281…282 °C (760 mmHg)
Melting Pt: 53…57 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004051
CAS Number: 580-13-2
EINECS: 209-452-5
Merck Index: 1301411

Order Now

Specification Test Results

Appearance White to light yellow to light beige Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 53 to 57 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR