1,4-Dibromonaphthalene 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
40650-0050
40650-0250
TS40650-0050EA
134
USD
TS40650-0050
TS40650-0250
1,4-Dibromonaphthalene 98+%
1,4-Dibromonaphthalene
Formula:
C₁₀H₆Br₂ Melting Pt: 80…82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041823 CAS Number: 83-53-4 EINECS: 201-484-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...