Tetrachlorocyclopropene 98%

Supplier: Thermo Scientific Chemicals
Warning

42019-0050 42019-0250
TS42019-0050EA 187 USD
TS42019-0050 TS42019-0250
Tetrachlorocyclopropene 98%
Tetrachlorocyclopropene
Formula: C₃Cl₄
Boiling Pt: 125…130 °C
Storage Temperature: Refrigerator
CAS Number: 6262-42-6
EINECS: 228-418-0

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5063 to 1.5083 (20°C, 589 nm)
GC ≥97.5 %
Stabilizer Approx. 0.01 % Citric acid
Relative density (20/4°C) 1.026 to 1.036
Acid value ≤5 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR