8-Chloroquinoline 99%

Supplier: Thermo Scientific Chemicals
Warning

40502-0100 40502-0250
TS40502-0100EA 62 USD
TS40502-0100 TS40502-0250
8-Chloroquinoline 99%
8-Chloroquinoline
Formula: C₉H₆ClN
Boiling Pt: 288 °C (1013 hPa)
Melting Pt: –20 °C
Flash Pt: >110 °C
MDL Number: MFCD00047618
CAS Number: 611-33-6
EINECS: 210-265-6

Order Now

Specification Test Results

Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
GC ≥98.5 %
Residue after evaporation ≤0.001 %
Free amine ≤0.0002 %
Acidity (CH3COOH) ≤0.003 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR