1,3-Dichlorobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

15118-0010 15118-0250 15118-2500 15118-5000
TS15118-0010EA 219 USD
TS15118-0010 TS15118-0250 TS15118-2500 TS15118-5000
1,3-Dichlorobenzene 98%
1,3-Dichlorobenzene
Formula: C₆H₄Cl₂
MW: 147 g/mol
Boiling Pt: 173
Melting Pt: -25
Density: 1.28 g/cm³
Flash Pt: 67 °C
Storage Temperature: Ambient
MDL Number: MFCD00000573
CAS Number: 541-73-1
EINECS: 208-792-1
UN: 3082
ADR: 9 III
Merck Index: 13,03080

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5450 to 1.5470 (20°C, 589 nm)
Water ≤0.1 %
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR