1-(2-Bromoethoxy)-4-chlorobenzene 99%

Supplier: Thermo Scientific Chemicals
Warning

37983-0050 37983-0250
TS37983-0050EA 133 USD
TS37983-0050 TS37983-0250
1-(2-Bromoethoxy)-4-chlorobenzene 99%
1-(2-Bromoethoxy)-4-chlorobenzene
Formula: C₈H₈BrClO
Boiling Pt: 150…152 °C (18 mmHg)
Melting Pt: 38…39 °C
Flash Pt: 150 °C
MDL Number: MFCD00000617
CAS Number: 2033-76-3
EINECS: 217-994-9
UN: 2811

Order Now

Specification Test Results

Appearance Clear liquid or low melting solid
Infrared spectrum Conforms
Refractive index 1.5570 to 1.5590 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR