3-Chloro-2-(chloromethyl)propene 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Chloro-2-chloromethyl-1-propene, 3-Chloro-2-(chloromethyl)-1-propene

27707-0050 27707-0250 27707-1000
TS27707-0050EA 95 USD
TS27707-0050 TS27707-0250 TS27707-1000
3-Chloro-2-(chloromethyl)propene 96%
3-Chloro-2-(chloromethyl)propene
Formula: C₄H₆Cl₂
MW: 125 g/mol
Boiling Pt: 138…140 °C (760 mmHg)
Melting Pt: 14 °C
Density: 1.08 g/cm³
Flash Pt: 36 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000952
CAS Number: 1871-57-4
EINECS: 217-489-3
UN: 2929
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4750 to 1.4790 (20°C, 589 nm)
GC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR