1-Bromo-3-methyl-2-butene 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3,3-Dimethylallyl bromide, 4-Bromo-2-methyl-2-butene

30741-0100 30741-0500
TS30741-0100EA 52 USD
TS30741-0100 TS30741-0500
1-Bromo-3-methyl-2-butene 96%
1-Bromo-3-methyl-2-butene
Formula: C₅H₉Br
MW: 149.03 g/mol
Boiling Pt: 82…83 °C (150 mmHg)
Density: 1.27 g/cm³
Flash Pt: 32 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000242
CAS Number: 870-63-3
EINECS: 212-799-5
UN: 2920
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.4890 to 1.4910 (20°C, 589 nm)
Titration Argentometric ≥95 % (ex Br)
Stabilizer silver wool

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR