5-Bromo-1-pentene 95%

Supplier: Thermo Scientific Chemicals
Warning

29517-0025 29517-0100 29517-0500
TS29517-0025EA 42 USD
TS29517-0025 TS29517-0100 TS29517-0500
5-Bromo-1-pentene 95%
5-Bromo-1-pentene
Formula: C₅H₉Br
Boiling Pt: 126…127 °C (765 mmHg)
Density: 1.26 g/cm³
Flash Pt: 30 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000264
CAS Number: 1119-51-3
EINECS: 214-281-4
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.4620 to 1.4650 (20°C, 589 nm)
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR