Neopentyl bromide 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,2-Dimethyl-1-propyl bromide, 1-Bromo-2,2-dimethylpropane

42572-0010 42572-0050 42572-0250
TS42572-0010EA 33 USD
TS42572-0010 TS42572-0050 TS42572-0250
Neopentyl bromide 98%
Neopentyl bromide
Formula: C₅H₁₁Br
MW: 151.05 g/mol
Boiling Pt: 105…106 °C (767 mmHg)
Density: 1.199 g/cm³
Flash Pt: 7 °C
Storage Temperature: Ambient
MDL Number: MFCD00000209
CAS Number: 630-17-1
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4350 to 1.4390 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR