1,2-Dibromo-2-methylpropane 98%

Supplier: Thermo Scientific Chemicals
Warning

40649-0050 40649-0250 40649-2500
TS40649-0050EA 32 USD
TS40649-0050 TS40649-0250 TS40649-2500
1,2-Dibromo-2-methylpropane 98%
1,2-Dibromo-2-methylpropane
Formula: C₄H₈Br₂
MW: 215.92 g/mol
Boiling Pt: 150 °C (760 mmHg)
Density: 1.79 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000126
CAS Number: 594-34-3
EINECS: 209-835-7

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5083 to 1.5103 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR