1-Iodododecane 98% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Dodecyl iodide
25009-0250
25009-1000
25009-5000
TS25009-0250EA
51
USD
TS25009-0250
TS25009-1000
TS25009-5000
1-Iodododecane 98% stabilized
1-Iodododecane
Formula:
C₁₂H₂₅I MW: 296.24 g/mol Boiling Pt: 159…160 °C (15 mmHg) Melting Pt: –3 °C Density: 1.2 g/cm³ Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001088 CAS Number: 4292-19-7 EINECS: 224-293-1 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4825 to 1.4865 (20°C, 589 nm) |
GC | ≥97.5 % |
Stabilizer | Copper |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...