1-Iodoperfluorohexane 97%

Supplier: Thermo Scientific Chemicals
Warning

26994-0050 26994-0250 26994-1000
TS26994-0050EA 39 USD
TS26994-0050 TS26994-0250 TS26994-1000
1-Iodoperfluorohexane 97%
1-Iodoperfluorohexane
Formula: C₆F₁₃I
Boiling Pt: 116…119 °C (760 mmHg)
Melting Pt: –45 °C
Density: 2.063 g/cm³
Flash Pt: >115 °C
Storage Temperature: Ambient
MDL Number: MFCD00001063
CAS Number: 355-43-1

Order Now

Specification Test Results

Appearance Clear light yellow to violet Liquid
Infrared spectrum Conforms
Refractive index 1.3275 to 1.3295 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR