1,4-Dichlorobutane 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
21643-0010
21643-1000
21643-5000
TS21643-0010EA
151
USD
TS21643-0010
TS21643-1000
TS21643-5000
1,4-Dichlorobutane 99%
1,4-Dichlorobutane
Formula:
ClH₂C(CH₂)₂CH₂Cl MW: 127.01 g/mol Boiling Pt: 153.9 Melting Pt: -38 Density: 1.16 g/cm³ Flash Pt: 40 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001011 CAS Number: 110-56-5 EINECS: 203-778-1 UN: 1993 ADR: 3 III |
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4530 to 1.4550 (20°C, 589 nm) |
Water | ≤0.05 % |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...