Pelargonic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Nonanoic acid, Pelargonic Acid

35624-0010 35624-0025 35624-1000 35624-5000
TS35624-0010EA 60 USD
TS35624-0010 TS35624-0025 TS35624-1000 TS35624-5000
Pelargonic acid 97%
Pelargonic acid
Formula: C₉H₁₈O₂
MW: 158.24 g/mol
Boiling Pt: 254 °C (760 mmHg)
Melting Pt: 12.5 °C
Density: 0.9 g/cm³
Flash Pt: 114 °C
Storage Temperature: Ambient
MDL Number: MFCD00004433
CAS Number: 112-05-0
EINECS: 203-931-2
Merck Index: 1207198

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4300 to 1.4340 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR