1-Bromoperfluorooctane 99%

Supplier: Thermo Scientific Chemicals
Warning

38679-0100 38679-0500
TS38679-0100EA 122 USD
TS38679-0100 TS38679-0500
1-Bromoperfluorooctane 99%
1-Bromoperfluorooctane
Formula: BrF₂C(CF₂)₆CF₃
Boiling Pt: 142 °C (760 mmHg)
Melting Pt: 6 °C
Density: 1.93 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00042082
CAS Number: 423-55-2
EINECS: 207-028-4
Merck Index: 1207299

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR