Taqdog Hot Start 2× Green Master Mix , Bulldog Bio
Supplier: Bulldog Bio
TDHM020 TDHM100
104016-444EA 49.5 USD
104016-444 104016-446 104016-448
Taqdog Hot Start 2× Green Master Mix , Bulldog Bio
Nucleic Acid Reagents End-point PCR Enzymes and Kits
TaqDog 2× Green Hot-Start Master Mixes provide everything needed to perform PCR. Just add template and primers and then start amplifying.
  • Faster preparation time
  • Enhanced for hard-to-amplify targets
  • Room temperature preparation
  • Loading dye for direct gel loading
  • Robust and stable for many months
TaqDog 2× Green Master Mixes provide everything needed to perform PCR. Just add template and primers and then start amplifying. These master mixes are based upon our popular TaqDog DNA Polymerases and includes nucleotides, buffer, and a brilliant green dye allowing for amplified samples to be directly loaded onto gels. Formulated for a 50 ul reaction size, its robust processivity makes smaller reaction volumes an economical option for labs looking to stretch their lab’s dollars. TaqDog master mixes come in two versions, premium and hot start. Our TaqDog Premium master mix is our most affordable option and provides great results in most cases. The TaqDog Hot Start master mix includes an aptamer-based hot start technology that promises better yields with no interference from primer-dimers.
Some PCR master mixes come with a separate tube of 'PCR enhancer' for more difficult templates, especially those with increased G-C nucleotide content. The additional hydrogen bonding for G-C pairs makes PCR less efficient — often resulting in weak results. Both types of TaqDog 2× Green Master Mixes also include their own unique enhancer reagents that can be added when amplification efficiency needs to be improved.
Hot start PCR has been around for over 20 years, and has been conclusively shown to reduce amplification artifacts caused by low-temperature 'false'-priming. Many methods have been developed over this time, but TaqDog Hot Start’s aptamer technology combines their best features while eliminating their worst drawbacks. It’s based upon a DNA construct engineered to form a secondary hairpin structure that reversibly-binds Taq at room temperature. When bound, the polymerase activity is inhibited at temperature at or below 45 °C. This prevents the extension of incorrectly primed regions in a sample’s template DNA. At higher temperatures, the aptamer falls off, and only strongly binding, and specific, primers can then initiate polymerization. This results in final PCR products that are clear of extra, unwanted bands, including the notoriously aggravating primer-dimer bands.
TaqDog Hot Start aptamers are synthetically prepared and HPLC purified for both a precise and defined composition which greatly reduces lot-to-lot variation. This also avoids contamination with trace amounts of mammalian DNA or cellular contents that can be associated with antibody preps. TaqDog Hot Start aptamers are inert and immune to proteases. They contain a 3′-cap that prevents their amplification during PCR or even ability to be cloned. Interference of aptamers with downstream applications has not been observed. Moreover, DNA aptamers do not become irreversibly denatured over time (due to high temperatures), but in contrast, allow for more robust amplification and higher yields. Unlike antibody-mediated inhibition, aptamers are able to re-bind and inhibit our polymerase after the completion of a PCR protocol. This is useful in applications where baseline signal is critical and even modest post-PCR activity (for example during long long-temperature 'soak' steps common to many protocols) can lead to artifacts in data.