American Machine & Tool Motor Foot Bracket
Supplier: American Machine & Tool
1506-000-00
MFLX07127-80EA 29 USD
MFLX07127-80
American Machine & Tool Motor Foot Bracket
Pumps Motors
Create your own motor mounting with the hardware below.
  • Easily mount your American Machine & Tool Motor NEMA Type 56 C-face motors
Motor Foot Bracket provides a base for NEMA Type 56 C-face motors.