Spun Polypropylene Progressive-Density Cartridge
Supplier: Avantor
MFLX01509-37EA 17.6 USD
MFLX01509-37 MFLX01509-41
Spun Polypropylene Progressive-Density Cartridge
Filters Filter Cartridges
Spun gradient-density construction provides greater dirt holding capacity.
  • Constructed of pleated polypropylene
Ideal for removing a broad range of deformable and nondeformable particles such as sand, silt, sludge, rust, and scale. Polypropylene construction is resistant to chemical and bacterial attack. Suitable to 145 °F (63 °C). Note: For optimal performance, change cartridge at 25 psid.