BS3 (Bis(sulfosuccinimidyl) suberate sodium salt) ≥95% (by NMR), Sigma-Aldrich®

Synonyms: bis(sulphosuccinimidyl)suberate, Suberic acid bis(3-sulpho-N-hydroxysuccinimide ester) sodium salt, BS³, Sulfo DSS

M-Clarity™ quality level = MQ300

S5799-100MG S5799-25MG
220209-750EA 407.08 USD
220209-750 220209-751
BS3 (Bis(sulfosuccinimidyl) suberate sodium salt) ≥95% (by NMR), Sigma-Aldrich®
BS3 (Bis(sulfosuccinimidyl) suberate sodium salt)

Suberic acid bis(3-sulfo-N-hydroxysuccinimide ester) sodium salt (BS3) is a water soluble, homobifunctional cross-linking reagent with amine reactivity. It is typically coupled to molecules containing primary amines by amide bonds buffered at pH 7.5 (6.5-8.5). It incorporates 8-atom linker.


Suberic acid bis(3-sulfo-N-hydroxysuccinimide ester) sodium salt (BS3) is useful for the preparation of protein-protein, receptor-ligand and hapten carrier molecule conjugates.

Formula: C₁₆H₁₈N₂Na₂O₁₄S₂
MW: 572.43 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00054982
CAS Number: 82436-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR