Uromodulin Human NATIVE (Human urine)
Supplier: BioVendor
RD172163100 RD172163025
102552-840EA 335.85 USD
102552-840 103739-800
Uromodulin Human NATIVE (Human urine)
Proteins and Peptides
Native protein isolated from human urine, 590 AA, 64265 kDa (calculated without glycosylation). Protein identity confirmed by LC-MS/MS.
  • Quality control test
  • BCA to determine quantity of the protein
  • SDS PAGE to determine purity of the protein
  • GFC to determine purity of the protein
  • LAL to determine quantity of endotoxin
Precaución: Research use only.