2,4,5,6-Tetraaminopyrimidine sulphate hydrate 98%

Supplier: Thermo Scientific Chemicals
Warning

13793-0050 13793-0250
TS13793-0050EA 14 USD
TS13793-0050 TS13793-0250
2,4,5,6-Tetraaminopyrimidine sulphate hydrate 98%
2,4,5,6-Tetraaminopyrimidine sulfate hydrate
Formula: C₄H₈N₆·H₂SO₄·xH₂O
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD06408008
CAS Number: 5392-28-9
EINECS: 226-393-0

Order Now

Specification Test Results

Appearance Ochre to brown Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤9 % (1 g, 150°C, 3 hrs)
Titration with NaOH ≥97.5 % (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR