5-Bromo-4-chloro-3-indolyl phosphate disodium salt hydrate (BCIP-Na hydrate) 98%

Supplier: Thermo Scientific Chemicals

27146-0010 27146-0500 27146-2500
TS27146-0010EA 424 USD
TS27146-0010 TS27146-0500 TS27146-2500
5-Bromo-4-chloro-3-indolyl phosphate disodium salt hydrate (BCIP-Na hydrate) 98%
5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na)
Formula: C₈H₄BrClNNa₂O₄P·xH₂O
MW: 370.43 g/mol (anhy)
Melting Pt: 300 °C
Storage Temperature: Freezer
MDL Number: MFCD00150468
CAS Number: 205926-98-3

Order Now

Specification Test Results

Appearance White to light beige to light bleu Powder
Infrared spectrum Conforms
Water 5 to 9 % (K.F.)
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR