3-Cyanophenylacetic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

43489-0010 43489-0050
TS43489-0010EA 151 USD
TS43489-0010 TS43489-0050
3-Cyanophenylacetic acid 95%
3-Cyanophenylacetic acid
Formula: C₉H₇NO₂
Melting Pt: 117…118 °C
Storage Temperature: Ambient
MDL Number: MFCD06411079
CAS Number: 1878-71-3
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to brown Powder
Infrared spectrum Conforms
Melting point 113 to 119 °C
HPLC ≥94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR