Isopropyl bromoacetate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: isopropyl 2-bromoacetate, Acetic acid, bromo-, 1-methylethyl ester, Acetic acid, 2-bromo-, 1-methylethyl ester

41281-0250 41281-1000
TS41281-0250EA 73 USD
TS41281-0250 TS41281-1000
Isopropyl bromoacetate 97%
Isopropyl bromoacetate
Formula: C₅H₉BrO₂
Boiling Pt: 59…61 °C (10 mmHg)
Density: 1.399 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00009886
CAS Number: 29921-57-1

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4440 to 1.4460 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR